Assay Details
Gene Symbol: TSACC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003355128.5 | XP_003355176.1 | 282 | 150 | ||
XM_013996979.2 | XP_013852433.1 | 304 | 150 | |||
XM_013996980.2 | XP_013852434.1 | 898 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
