Assay Details
Gene Symbol: TSACC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013996979.2 | XP_013852433.1 | 126 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013996979.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
