Assay Details
Gene Symbol: RASSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021091944.1 | XP_020947603.1 | 1579 | 101 | ||
XM_021091945.1 | XP_020947604.1 | 326 | 101 | |||
XM_021091946.1 | XP_020947605.1 | 371 | 101 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
