Assay Details
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005653133.3 | XP_005653190.1 | 1273 | 72 | ||
XM_005653134.3 | XP_005653191.1 | 1240 | 72 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
