Assay Details
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356107.4 | XP_003356155.1 | 63 | 117 | ||
XM_005653340.3 | XP_005653397.1 | 38 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003356107.4, XM_005653340.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
