Assay Details
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003356855.5 | XP_003356903.1 | 450 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
