Assay Details
Gene Symbol: CMPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021087907.1 | XP_020943566.1 | 1483 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
