Assay Details
Gene Symbol: DCUN1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021086515.1 | XP_020942174.1 | 188 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021086515.1 transcripts.
Set Membership: |
![]() ![]() |
