Assay Details
Gene Symbol: AKAP17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021081363.1 | XP_020937022.1 | 159 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021081363.1 transcripts.
Set Membership: |
![]() ![]() |
