Assay Details
Gene Symbol: GPR153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021095249.1 | XP_020950908.1 | 1330 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
