Assay Details
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK345924.1 | - | 2 | 227 | 109 | |
AK395844.1 | - | 2 | 253 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SRC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK345924.1 | - | 2 | 227 | 109 | |
AK395844.1 | - | 2 | 253 | 109 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |