Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001928850.6 | XP_001928885.1 | 3045 | 106 | ||
XM_005660498.3 | XP_005660555.1 | 3372 | 106 | |||
XM_005660499.3 | XP_005660556.1 | 2917 | 106 | |||
XM_013993913.2 | XP_013849367.1 | 3452 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
