Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244313.1 | NP_001231242.1 | 11 - 12 | 1020 | 109 | |
XM_021076560.1 | XP_020932219.1 | 677 | 109 | |||
GenBank mRNA | AK232580.1 | - | 11 - 12 | 1022 | 109 | |
AY609935.1 | - | 10 - 11 | 920 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
