Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244313.1 | NP_001231242.1 | 1 - 2 | 203 | 54 | |
GenBank mRNA | AK232580.1 | - | 1 - 2 | 205 | 54 | |
AK343903.1 | - | 1 - 2 | 203 | 54 | ||
AK345134.1 | - | 1 - 2 | 229 | 54 | ||
AY609935.1 | - | 1 - 2 | 168 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
