Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244313.1 | NP_001231242.1 | 3 - 4 | 364 | 83 | |
GenBank mRNA | AK232580.1 | - | 3 - 4 | 366 | 83 | |
AK343903.1 | - | 3 - 4 | 364 | 83 | ||
AK345134.1 | - | 3 - 4 | 390 | 83 | ||
AY609935.1 | - | 3 - 4 | 329 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
