Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244313.1 | NP_001231242.1 | 4 - 5 | 427 | 69 | |
XM_021076560.1 | XP_020932219.1 | 84 | 69 | |||
GenBank mRNA | AK232580.1 | - | 4 - 5 | 429 | 69 | |
AK343903.1 | - | 4 - 5 | 427 | 69 | ||
AK345134.1 | - | 4 - 5 | 453 | 69 | ||
AY609935.1 | - | 4 - 5 | 392 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021076560.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
