Assay Details
Gene Symbol: DIMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244313.1 | NP_001231242.1 | 9 - 10 | 850 | 117 | |
XM_021076560.1 | XP_020932219.1 | 507 | 117 | |||
GenBank mRNA | AK232580.1 | - | 9 - 10 | 852 | 117 | |
AK343903.1 | - | 9 - 10 | 850 | 117 | ||
AK345134.1 | - | 9 - 10 | 876 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
