Assay Details
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244547.1 | NP_001231476.1 | 6 - 7 | 1004 | 109 | |
GenBank mRNA | AK345125.1 | - | 6 - 7 | 1004 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
