Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 16 - 17 | 1819 | 72 | |
XM_021066120.1 | XP_020921779.1 | 2184 | 72 | |||
XM_021066122.1 | XP_020921781.1 | 2360 | 72 | |||
XM_021066123.1 | XP_020921782.1 | 1806 | 72 | |||
XM_021066124.1 | XP_020921783.1 | 529 | 72 | |||
XM_021066125.1 | XP_020921784.1 | 2934 | 72 | |||
GenBank mRNA | DN102622.1 | - | 3 - 4 | 366 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
