Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 17 - 18 | 1938 | 89 | |
XM_021066120.1 | XP_020921779.1 | 2303 | 89 | |||
XM_021066122.1 | XP_020921781.1 | 2479 | 89 | |||
XM_021066123.1 | XP_020921782.1 | 1925 | 89 | |||
XM_021066124.1 | XP_020921783.1 | 648 | 89 | |||
XM_021066125.1 | XP_020921784.1 | 3053 | 89 | |||
GenBank mRNA | DN102622.1 | - | 2 - 3 | 247 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
