Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 18 - 19 | 2067 | 131 | |
XM_021066120.1 | XP_020921779.1 | 2432 | 131 | |||
XM_021066122.1 | XP_020921781.1 | 2608 | 131 | |||
XM_021066123.1 | XP_020921782.1 | 2054 | 131 | |||
XM_021066124.1 | XP_020921783.1 | 777 | 131 | |||
XM_021066125.1 | XP_020921784.1 | 3182 | 131 | |||
GenBank mRNA | DN102622.1 | - | 1 - 2 | 118 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
