Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 20 - 21 | 2312 | 87 | |
XM_021066120.1 | XP_020921779.1 | 2677 | 87 | |||
XM_021066122.1 | XP_020921781.1 | 2853 | 87 | |||
XM_021066123.1 | XP_020921782.1 | 2299 | 87 | |||
XM_021066124.1 | XP_020921783.1 | 1022 | 87 | |||
XM_021066125.1 | XP_020921784.1 | 3427 | 87 | |||
GenBank mRNA | DY419263.1 | - | 3 - 4 | 269 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
