Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 21 - 22 | 2458 | 63 | |
XM_021066120.1 | XP_020921779.1 | 2823 | 63 | |||
XM_021066122.1 | XP_020921781.1 | 2999 | 63 | |||
XM_021066123.1 | XP_020921782.1 | 2445 | 63 | |||
XM_021066124.1 | XP_020921783.1 | 1168 | 63 | |||
XM_021066125.1 | XP_020921784.1 | 3573 | 63 | |||
GenBank mRNA | DY419263.1 | - | 4 - 5 | 415 | 63 | |
FS705240.1 | - | 1 - 2 | 69 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
