Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 5 - 6 | 674 | 94 | |
XM_021066120.1 | XP_020921779.1 | 1039 | 94 | |||
XM_021066122.1 | XP_020921781.1 | 1215 | 94 | |||
XM_021066123.1 | XP_020921782.1 | 661 | 94 | |||
XM_021066125.1 | XP_020921784.1 | 1789 | 94 | |||
GenBank mRNA | DN102287.1 | - | 2 - 3 | 182 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
