Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253818.1 | NP_001240747.1 | 8 - 9 | 989 | 107 | |
XM_021066120.1 | XP_020921779.1 | 1354 | 107 | |||
XM_021066122.1 | XP_020921781.1 | 1530 | 107 | |||
XM_021066123.1 | XP_020921782.1 | 976 | 107 | |||
XM_021066125.1 | XP_020921784.1 | 2104 | 107 | |||
GenBank mRNA | DN102287.1 | - | 5 - 6 | 497 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
