Assay Details
Gene Symbol: FOSL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001315740.1 | NP_001302669.1 | 1 - 2 | 160 | 85 | |
GenBank mRNA | AK349073.1 | - | 1 - 2 | 446 | 85 | |
KM191330.1 | - | 1 - 2 | 160 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
