Assay Details
Gene Symbol: GLTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213822.2 | NP_998987.2 | 3 - 4 | 383 | 84 | |
GenBank mRNA | AK393749.1 | - | 3 - 4 | 383 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
