Assay Details
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214174.2 | NP_999339.2 | 1 - 2 | 198 | 130 | |
GenBank mRNA | AK239443.1 | - | 1 - 2 | 338 | 130 | |
AK346037.1 | - | 1 - 2 | 162 | 130 | ||
AK396895.1 | - | 1 - 2 | 197 | 130 | ||
AK399867.1 | - | 1 - 2 | 199 | 130 | ||
AY609423.1 | - | 1 - 2 | 180 | 130 | ||
BP142411.1 | - | 1 - 2 | 269 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
