Assay Details
Gene Symbol: CRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137636.2 | NP_001131108.1 | 1 - 2 | 356 | 99 | |
XM_021066038.1 | XP_020921697.1 | 405 | 99 | |||
GenBank mRNA | AK349082.1 | - | 1 - 2 | 392 | 99 | |
FJ175657.1 | - | 1 - 2 | 358 | 99 | ||
FJ175658.1 | - | 1 - 2 | 357 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
