Assay Details
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144902.1 | NP_001138374.1 | 2 - 3 | 250 | 75 | |
GenBank mRNA | AK392419.1 | - | 2 - 3 | 218 | 75 | |
AY609846.1 | - | 2 - 3 | 250 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
