Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159305.2 | NP_001152777.1 | 3 - 4 | 285 | 77 | |
XM_021090968.1 | XP_020946627.1 | 465 | 77 | |||
XM_021090969.1 | XP_020946628.1 | 489 | 77 | |||
XM_021090972.1 | XP_020946631.1 | 348 | 77 | |||
XM_021090973.1 | XP_020946632.1 | 314 | 77 | |||
XM_021090974.1 | XP_020946633.1 | 253 | 77 | |||
GenBank mRNA | AK235481.1 | - | 3 - 4 | 301 | 77 | |
AK238428.1 | - | 3 - 4 | 319 | 77 | ||
AK391628.1 | - | 3 - 4 | 317 | 77 | ||
FJ775535.1 | - | 3 - 4 | 286 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
