Assay Details
Gene Symbol: ATOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167641.2 | NP_001161113.1 | 3 - 4 | 360 | 131 | |
GenBank mRNA | AK234519.1 | - | 3 - 4 | 311 | 131 | |
AK391717.1 | - | 3 - 4 | 294 | 131 | ||
AK399932.1 | - | 3 - 4 | 362 | 131 | ||
AY610278.1 | - | 3 - 4 | 275 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
