Assay Details
Gene Symbol: MFSD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243400.1 | NP_001230329.1 | 1 - 2 | 337 | 103 | |
GenBank mRNA | AK237613.1 | - | 1 - 2 | 337 | 103 | |
AK396927.1 | - | 1 - 2 | 381 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
