Assay Details
Gene Symbol: ID3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243602.1 | NP_001230531.1 | 2 - 3 | 475 | 76 | |
GenBank mRNA | AK235079.1 | - | 2 - 3 | 450 | 76 | |
AK240309.1 | - | 2 - 3 | 475 | 76 | ||
AK394442.1 | - | 2 - 3 | 455 | 76 | ||
AY610252.1 | - | 2 - 3 | 431 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
