siRNA Details
Gene Symbol: Dcaf12l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190718.1 | NP_001177647.1 | 1 | 945 | ||
NM_178739.6 | NP_848854.3 | 1 | 981 | |||
GenBank mRNA | AK034343.1 | - | 1 | 798 | ||
AK034472.1 | - | 1 | 981 | |||
AK038557.1 | - | 1 | 936 | |||
AK039767.1 | - | 1 | 802 | |||
AK047360.1 | - | 1 | 945 | |||
BC068319.1 | - | 1 | 785 | 30431243 |
Gene Symbol: Dcaf12l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175539.3 | NP_780748.1 | 1 | 934 | ||
GenBank mRNA | AK037333.1 | - | 2 | 884 | ||
AK076436.1 | - | 941 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
