siRNA Details
Gene Symbol: DNAJA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK298725.1 | - | 2 | 199 | ||
BC008182.1 | - | 2 | 184 | 2900892 | ||
BT007292.1 | - | 1 | 80 | |||
D13388.1 | - | 2 | 197 | |||
L08069.1 | - | 2 | 162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
