siRNA Details
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178759.4 | NP_848874.3 | 9 | 1197 | ||
GenBank mRNA | AK046560.1 | - | 9 | 1196 | ||
AK079787.1 | - | 9 | 1198 | |||
AK080042.1 | - | 9 | 1196 | |||
AK080212.1 | - | 9 | 1198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
