siRNA Details
Gene Symbol: Sybu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285840.1 | NP_001272769.1 | 2 | 513 | ||
NM_176998.4 | NP_795972.2 | 3 | 714 | |||
NM_178765.3 | NP_848880.2 | 3 | 611 | |||
GenBank mRNA | AB232447.1 | - | 3 | 714 | ||
AB232449.1 | - | 3 | 611 | |||
AK029072.1 | - | 2 | 210 | |||
AK030482.1 | - | 3 | 343 | |||
AK030720.1 | - | 3 | 343 | |||
AK163432.1 | - | 3 | 376 | |||
AK220268.1 | - | 2 | 513 | |||
BC048945.1 | - | 2 | 337 | 6467420 | ||
BC065045.1 | - | 3 | 608 | 5686153 |

More Information
Other Species Targeted

Gene Ontology Categories:
