siRNA Details
Gene Symbol: Fry | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079097.1 | - | 10, 11 | 1316 | ||
AK220506.1 | - | 16, 17 | 2463 | |||
BC096532.1 | - | 15, 16 | 1978 |

More Information
Other Species Targeted

