siRNA Details
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178785.3 | NP_848900.2 | 2 | 645 | ||
GenBank mRNA | AK030797.1 | - | 2 | 648 | ||
AK037914.1 | - | 2 | 645 | |||
AK040573.1 | - | 2 | 633 | |||
AK041479.1 | - | 3 | 503 | |||
AK088449.1 | - | 2 | 607 | |||
AK088987.1 | - | 2 | 623 | |||
AK131128.1 | - | 4 | 552 | |||
BC132341.1 | - | 4 | 551 | 40130618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
