siRNA Details
Gene Symbol: Tmem117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178789.4 | NP_848904.1 | 2 | 333 | ||
GenBank mRNA | AK034024.1 | - | 2 | 287 | ||
AK047437.1 | - | 2 | 334 | |||
AK048597.1 | - | 2 | 293 | |||
BC051178.1 | - | 2 | 268 | 6314268 | ||
BC054070.1 | - | 2 | 268 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001271239.
Gene Ontology Categories:
