siRNA Details
Gene Symbol: Son | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178880.4 | NP_849211.3 | 9 | 7134 | ||
GenBank mRNA | AK034390.1 | - | 1 | 2830 | ||
AK035150.1 | - | 2 | 2625 | |||
AK122418.1 | - | 8 | 4250 | |||
BC145121.1 | - | 9 | 7074 | |||
CB849818.2 | - | 2 | 232 |

More Information
Other Species Targeted

Gene Ontology Categories:
