siRNA Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081264.1 | NP_001074733.1 | 3 | 331 | ||
GenBank mRNA | AK036792.1 | - | 3 | 293 | ||
AK087075.1 | - | 3 | 298 | |||
AK087630.1 | - | 3 | 290 | |||
AK157903.1 | - | 3 | 293 | |||
AK171887.1 | - | 3 | 284 | |||
BC050854.1 | - | 3 | 296 | 6332345 | ||
BC080867.1 | - | 3 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
