siRNA Details
Gene Symbol: Rasal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178785.3 | NP_848900.2 | 4, 5 | 800 | ||
GenBank mRNA | AK030797.1 | - | 4, 5 | 803 | ||
AK040573.1 | - | 4, 5 | 788 | |||
AK041479.1 | - | 5, 6 | 658 | |||
AK088449.1 | - | 4, 5 | 762 | |||
AK088987.1 | - | 4, 5 | 778 | |||
AK131128.1 | - | 6, 7 | 707 | |||
BC132341.1 | - | 6, 7 | 706 | 40130618 |

More Information
Other Species Targeted

Gene Ontology Categories:
