siRNA Details
Gene Symbol: Far2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178797.3 | NP_848912.1 | 2 | 301 | ||
GenBank mRNA | AK038584.1 | - | 2 | 254 | ||
AK043388.1 | - | 2 | 301 | |||
AK043589.1 | - | 2 | 301 | |||
AK137298.1 | - | 2 | 301 | |||
AK161201.1 | - | 2 | 268 | |||
BC055759.1 | - | 2 | 279 | 6809131 |
