siRNA Details
Gene Symbol: Prmt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081240.3 | NP_001074709.1 | 3 | 516 | ||
GenBank mRNA | AK045296.1 | - | 3 | 507 | ||
AK045315.1 | - | 3 | 506 | |||
AK154546.1 | - | 3 | 460 | |||
AK161023.1 | - | 3 | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
