siRNA Details
Gene Symbol: Timd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178759.4 | NP_848874.3 | 9 | 1958 | ||
GenBank mRNA | AK080042.1 | - | 9 | 1959 | ||
AY376716.1 | - | 9 | 1956 | |||
AY376717.1 | - | 9 | 1956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
