siRNA Details
Gene Symbol: Zrsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178794.4 | NP_848909.2 | 5 | 2032 | ||
GenBank mRNA | AK030296.1 | - | 5 | 1964 | ||
BC065693.1 | - | 5 | 1948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
