siRNA Details
Gene Symbol: Son | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178880.4 | NP_849211.3 | 12 | 7747 | ||
GenBank mRNA | AK035150.1 | - | 5 | 3238 | ||
AK122418.1 | - | 11 | 4863 | |||
BC029990.1 | - | 2 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
