siRNA Details
Gene Symbol: Smad6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008542.3 | NP_032568.3 | 4 | 2102 | ||
GenBank mRNA | AB221689.1 | - | 4 | 1067 | ||
AF010133.1 | - | 4 | 1067 | |||
AK004671.1 | - | 4 | 548 | |||
AK046602.1 | - | 4 | 2105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
